Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
I1905-28M
USBII1905-28MEA 8300 NOK
USBII1905-28M
Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgM (IgM)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR