Anti-JAK1 Rabbit Polyclonal Antibody

Leverandør: US Biological
J0900-02A
USBIJ0900-02AEA 4825 NOK
USBIJ0900-02A
Anti-JAK1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: JAK1 (Janus kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR