Anti-JAK1 Rabbit Polyclonal Antibody

Leverandør: US Biological
J0900-20B1
USBIJ0900-20B1EA 11350 NOK
USBIJ0900-20B1
Anti-JAK1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot:1:1000

Type: Primary
Antigen: JAK1 (Janus kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR