Anti-JAK2 Rabbit Polyclonal Antibody

Leverandør: US Biological
J0900-28C
USBIJ0900-28CEA 4675 NOK
USBIJ0900-28C
Anti-JAK2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: JAK2 (Janus kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR