Anti-JAK2 Goat Polyclonal Antibody

Leverandør: US Biological
J0901-04A
USBIJ0901-04AEA 10700 NOK
USBIJ0901-04A
Anti-JAK2 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 25 ?g/ml

Type: Primary
Antigen: JAK2 (Janus kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR