Anti-JAK3 Rabbit Polyclonal Antibody

Leverandør: US Biological
J0902-06
USBIJ0902-06EA 6675 NOK
USBIJ0902-06
Anti-JAK3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: JAK3 (Janus kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR