Anti-JAK3 Rabbit Monoclonal Antibody [clone: 10g85]

Leverandør: US Biological
J0902-06J
USBIJ0902-06JEA 9125 NOK
USBIJ0902-06J
Anti-JAK3 Rabbit Monoclonal Antibody [clone: 10g85]
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: JAK3 (Janus kinase 3)
Clonality: monoclonal
Clone: 10g85
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol JAK3
Antigen-navn Janus kinase 3
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon 10g85
Reaktivitet Human,Mouse
Vert Rabbit
Isotype IgG
Western blot Yes
Modifikasjon phospho-Tyr980, phospho-Tyr981
Format Supernatant
Skjema liquid
Lagringsbuffer 10mMsodium HEPES pH 7,5, 150mM NaCl, 0,1mg/ml BSA, 0,02% sodium azide, 50% glycerol.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR