Anti-KLH Mouse Monoclonal Antibody [clone: 9L437]

Leverandør: US Biological
K0300-02A
USBIK0300-02AEA 6225 NOK
USBIK0300-02A
Anti-KLH Mouse Monoclonal Antibody [clone: 9L437]
Antistoffer
Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: monoclonal
Clone: 9L437
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR