Anti-KLH Chicken Polyclonal Antibody

Leverandør: US Biological
K0300-02C
USBIK0300-02CEA 5225 NOK
USBIK0300-02C
Anti-KLH Chicken Polyclonal Antibody
Antistoffer
Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR