Anti-KLH Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
K0300-06
USBIK0300-06EA 11100 NOK
USBIK0300-06
Anti-KLH Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 1:20000-1:80000

Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR