Anti-KLH Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: US Biological
K0300-07
USBIK0300-07EA 11100 NOK
USBIK0300-07
Anti-KLH Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Recommended dilution: Immunofluorescence: 1:500-1:2500

Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR