Anti-KLH Chicken Polyclonal Antibody (FITC (Fluorescein))

Leverandør: US Biological
K0300-07A
USBIK0300-07AEA 5475 NOK
USBIK0300-07A
Anti-KLH Chicken Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR