Anti-LTF Mouse Monoclonal Antibody (Rhodamine) [clone: NI 25]

Leverandør: US Biological
L1021-83R
USBIL1021-83REA 10625 NOK
USBIL1021-83R
Anti-LTF Mouse Monoclonal Antibody (Rhodamine) [clone: NI 25]
Antistoffer
Recommended dilution: Immunochemistry: 1:10-1:100

Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: monoclonal
Clone: NI 25
Conjugation: Rhodamine
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR