Anti-Munc13-2 Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
M2196-71-BIOT
USBIM2196-71-BIOTEA 10875 NOK
USBIM2196-71-BIOT
Anti-Munc13-2 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Munc13-2
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR