Anti-Munc13-2 Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
M2196-71A-BIOT
USBIM2196-71A-BIOTEA 10875 NOK
USBIM2196-71A-BIOT
Anti-Munc13-2 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Munc13-2
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol MUNC13-2
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Biotin
Reaktivitet Human
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitop internal
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR