Anti-5-methyl-Cytidine Mouse Monoclonal Antibody [clone: 10B11]

Leverandør: US Biological
M3398-55M
USBIM3398-55MEA 11175 NOK
USBIM3398-55M
Anti-5-methyl-Cytidine Mouse Monoclonal Antibody [clone: 10B11]
Antistoffer
Type: Primary
Antigen: 5-Methylcytidine
Clonality: monoclonal
Clone: 10B11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR