Anti-Myokinase Sheep Polyclonal Antibody (Biotin)

Leverandør: US Biological
M9835-11B
USBIM9835-11BEA 8300 NOK
USBIM9835-11B
Anti-Myokinase Sheep Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Myokinase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR