Anti-NMS Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
N2171-79U2
USBIN2171-79U2EA 14275 NOK
USBIN2171-79U2
Anti-NMS Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: NMS (neuromedin S)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR