Anti-Neuropeptide B-29 Rabbit Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
N2175-91C5
USBIN2175-91C5EA 14275 NOK
USBIN2175-91C5
Anti-Neuropeptide B-29 Rabbit Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: Neuropeptide B-29
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR