Anti-Norfloxacin Sheep Polyclonal Antibody

Leverandør: US Biological
N4000-10B
USBIN4000-10BEA 12550 NOK
USBIN4000-10B
Anti-Norfloxacin Sheep Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Norfloxacin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigen-navn Norfloxacin
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Vert Sheep
Isotype IgG
ELISA Yes
Skjema liquid
Rensing purified
Konsentrasjon ~1 mg/ml
Lagringsbuffer 20mMphosphate, 150mM sodium chloride, pH 7,2, 0,09% sodium azide.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR