Anti-Nucleoside Monophosphate Kinase Rabbit Polyclonal Antibody

Leverandør: US Biological
N7100-69A
USBIN7100-69AEA 10050 NOK
USBIN7100-69A
Anti-Nucleoside Monophosphate Kinase Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Nucleoside Monophosphate Kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR