Osteoclast inhibitory lectin MAB MO XMO
Leverandør: US Biological
O8061-100
USBIO8061-100EA 7725 NOK
USBIO8061-100
Osteoclast inhibitory lectin MAB MO XMO
Assay Proteinanalyser
Osteocrin is a recently identified secreted protein expression of which was only detected in bone, peaking just after birth and decreasing markedly with age. A 1280-bp mRNA encodes osteocrin producing a mature protein of 103 amino acids with a molecular mass of 11,4 kD. In primary osteoblastic cell cultures osteocrin expression coincided with matrix formation then decreased in very mature cultures.
Treatment of cultures with 1,25-dihydroxyvitamin D3 resulted in a rapid dose-dependent down-regulation of osteocrin expression, suggesting direct regulation. Chronic treatment of primary cultures with osteocrin-conditioned media inhibited mineralization and reduced osteocalcin and alkaline phosphatase expression. These results suggest that osteocrin represents a novel, unique vitamin D-regulated bone-specific protein that appears to act as a soluble osteoblast regulator.