Anti-OSTN Rabbit Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
O8061-18L5
USBIO8061-18L5EA 14275 NOK
USBIO8061-18L5
Anti-OSTN Rabbit Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: OSTN (osteocrin)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR