Anti-NAMPT Rabbit polyclonal antibody Rhodamine

Leverandør: US Biological
P3113-96Z5
USBIP3113-96Z5EA 14275 NOK
USBIP3113-96Z5
Anti-NAMPT Rabbit polyclonal antibody Rhodamine
Antistoffer
Type: Primary
Antigen: NAMPT (nicotinamide phosphoribosyltransferase)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR