Anti-Phosphodiesterase I Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
P4071-97B
USBIP4071-97BEA 7700 NOK
USBIP4071-97B
Anti-Phosphodiesterase I Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Phosphodiesterase I
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigen-navn Phosphodiesterase I
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Biotin
Reaktivitet
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Skjema lyophilized powder
Rensing purified by ammonium sulphate precipitation and ion exchange chromatography
Lagringsbuffer PBS, pH 7,2. No
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR