Anti-PROK1 Rabbit polyclonal antibody Rhodamine

Leverandør: US Biological
P9008-80J
USBIP9008-80JEA 14275 NOK
USBIP9008-80J
Anti-PROK1 Rabbit polyclonal antibody Rhodamine
Antistoffer
Type: Primary
Antigen: PROK1 (prokineticin 1)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR