Anti-PXK Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: US Biological
P9395-01A-AP
USBIP9395-01A-APEA 10875 NOK
USBIP9395-01A-AP
Anti-PXK Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: PXK (PX domain containing serine/threonine kinase)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol PXK
Antigen-navn PX domain containing serine/threonine kinase
Antistofftype Primary
Klonalitet Polyclonal
Konjugering AP (Alkaline Phosphatase)
Reaktivitet Human
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitop internal
Skjema liquid
Rensing purified by Protein G chromatography
Lagringsbuffer PBS, pH 7,2, contains ~0,09% sodium azide
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR