Anti-Pyroglutamate Aminopeptidase Rabbit Polyclonal Antibody

Leverandør: US Biological
P9539-90A
USBIP9539-90AEA 9950 NOK
USBIP9539-90A
Anti-Pyroglutamate Aminopeptidase Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Pyroglutamate Aminopeptidase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR