Anti-Rhodamine Goat Polyclonal Antibody

Leverandør: US Biological
R1998-12J
USBIR1998-12JEA 4300 NOK
USBIR1998-12J
Anti-Rhodamine Goat Polyclonal Antibody
Antistoffer
Type: Tag
Antigen: Rhodamine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol Rhodamine
Environmentally Preferable
Antistofftype Tag
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Goat
Isotype IgG
ELISA Yes
Skjema liquid
Rensing purified by affinity chromatography
Konsentrasjon ~1 mg/ml
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR