Anti-SH3 and PX domain-containing protein 2B Rabbit Polyclonal Antibody

Leverandør: US Biological
S1013-33A
USBIS1013-33AEA 7625 NOK
USBIS1013-33A
Anti-SH3 and PX domain-containing protein 2B Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 3?g/ml; Immunochemistry: 1:3000

Type: Primary
Antigen: SH3 and PX domain-containing protein 2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

SPESIFIKASJONER

Antigen-navn SH3 and PX domain-containing Protein 2B
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Mouse
Vert Rabbit
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Immunutfelling Yes
Aminosyretall 257 to 477
Skjema liquid
Rensing purified by Protein A chromatography
Konsentrasjon ~1 mg/ml
Lagringsbuffer Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR