Anti-SH3 and PX domain-containing protein 2B Rabbit Polyclonal Antibody

Leverandør: US Biological
S1013-33B
USBIS1013-33BEA 7150 NOK
USBIS1013-33B
Anti-SH3 and PX domain-containing protein 2B Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 3?g/ml

Type: Primary
Antigen: SH3 and PX domain-containing protein 2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestill nå

SPESIFIKASJONER

Antigen-navn SH3 and PX domain-containing Protein 2B
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Mouse
Vert Rabbit
Western blot Yes
Immunutfelling Yes
Aminosyretall 431 to 595
Skjema liquid
Rensing purified by Protein A chromatography
Konsentrasjon ~1 mg/ml
Lagringsbuffer Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR