Anti-SORL1 Rabbit Polyclonal Antibody (Rhodamine)

Leverandør: US Biological
S5353-02J
USBIS5353-02JEA 10275 NOK
USBIS5353-02J
Anti-SORL1 Rabbit Polyclonal Antibody (Rhodamine)
Antistoffer
Type: Primary
Antigen: SORL1 (sortilin-related receptor, L(DLR class) A repeats containing)
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR