Anti-SULT4A1b Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
S8022-04A-BIOT
USBIS8022-04A-BIOTEA 10875 NOK
USBIS8022-04A-BIOT
Anti-SULT4A1b Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: SULT4A1b
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR