Disposable earplugs, uvex x-fit detec
Leverandør: UVEX
2112.011
UVEX2112.011EA 2050 NOK
UVEX2112.011
Disposable earplugs, uvex x-fit detec
Ørepropper Ørepropper til engangsbruk
The x-fit detect is a detectable disposable earplug for the food industry. The connecting cord makes it practical to wear and limits the risk of process contamination.
  • Ergonomic shape adapts to the natural shape of the ear canal
  • 'x' emboss in the end of the plug makes it easy to remove while simultaneously reducing the pressure on the outer wall of the ear canal
  • Easy insertion and removal
  • For medium to large ear canals
  • Bright blue colour for easy recognition in production areas
SNR: 37 dB
Sertifiseringer: EN 352-2:2002