Leverandør: UVEX
8763220 8763310 8763110 8763210 8763200 8763100
UVEX8763220EA 690 NOK
UVEX8763220 UVEX8763310 UVEX8763110 UVEX8763210 UVEX8763200 UVEX8763100
Respiratorer Engangsmasker Støvmasker
Respirators in protection class FFP1, FFP2 and FFP3.
  • Soft material edges for greater comfort and wearer acceptance
  • Continuous textile headband ensures that the mask fits comfortably and securely
  • Adjustable nose clip ensures an excellent individual fit and secure positioning
  • Optional: exhalation valve to reduce heat and humidity in the mask
More sustainable mask family: the valve and headband attachment on all models are partly made of recycled material.
The soft material edges prevent uncomfortable pressure points. The seamless headband can beMore Product Information