Uvex Planet Safety Spectacle
Leverandør: UVEX
9128295
UVEX9128295EA 355 NOK
UVEX9128295
Uvex Planet Safety Spectacle
Vernebriller
Attractive, classic, sporty design, pleasantly lightweight and optimal protection - the uvex pheos nxt. The special design of the arms not only looks impressive: the direction-focused structure also provides extremely good grip - even during demanding tasks. Numerous openings in the side arms improve air circulation and make the glasses light yet no less robust. The large panoramic lens also provides a wide field of vision free from distortion to ensure a good view.
  • Classic, sporty and lightweight with innovative arm design for an optimal, reliable fit