Goggles, ultravision wide-vision gas-tight
Leverandør: UVEX
9301.613
UVEX9301.613EA 470 NOK
UVEX9301.613
Goggles, ultravision wide-vision gas-tight
Vernebriller /Goggles
Classic, ergonomic wide-vision goggle with unrestricted side visibility.
  • Anti-fog coating on the inside, extremely resistant to scratches and chemicals on the outside
  • Gas-tight version without a ventilation system
  • Large goggle body, can also comfortably be worn as overspecs
  • Yellow frame
  • Clear PC lens
The uvex ultravision features a comfortable and ergonomic design and offers unrestricted side visibility. This flexible wide-vision goggle is also suitable for use as overspecs. The uvex supravision excellence coating provides anti-fog protection on theMore Product Information