Goggles, uvex 9305
Leverandør: UVEX
9305.714
UVEX9305.714EA 550 NOK
UVEX9305.714
Goggles, uvex 9305
Vernebriller /Goggles
Classic safety goggles with acetate lens.
  • Effective ventilation system
  • Adjustable textile headband
  • Mechanical strength: F –Low energy impact protection (45 m/s)
Sertifiseringer: EN 166, EN 170