Safety helmets, uvex pheos B
Leverandør: UVEX
9772.120 9772.020 9772.220 9772.320 9772.520 9772.920
UVEX9772.120EA 235 NOK
UVEX9772.120 UVEX9772.020 UVEX9772.220 UVEX9772.320 UVEX9772.520 UVEX9772.920
Safety helmets, uvex pheos B
Hjelmer Sikkerhethjelm
These ventilated helmets provide protection even in extremely low temperatures (down to −30 °C). They also provide protection against splashes of molten metal.
  • Three variable air vents aids airflow and helps regulate the temperature within the helmet making it more comfortable to wear
  • Adapter systems for attaching uvex visors, uvex earmuffs and lamps
  • Suspension harness for comfortable adjustable fit
Size: 520 - 610 mm
Sertifiseringer: EN 397, low temperature (–30 °C), and molten metal