Safety helmet, pheos S-KR
Leverandør: UVEX
9772.544
UVEX9772.544EA 570 NOK
UVEX9772.544
Safety helmet, pheos S-KR
Hjelmer Sikkerhethjelm
This safety helmet is equipped with slots for uvex visors and earmuffs.
  • Shorter brim for a wider field of upward vision
  • Three variable air vents for maximum ventilation
  • Adjustable range for helmet is 520 to 610 mm
  • Optimum fit and comfort with six point textile suspension
The suspension harness features a wheel ratchet system for individual adjustment, with the possibility to mount uvex pheos goggles. Helmet is premounted with four point chinstrap and a goggle glip.
Adjustable range: 520 to 610 mm