Magnetic Wall Bracket
Leverandør: UVEX
9963.004
UVEX9963.004EA 375 NOK
UVEX9963.004
Magnetic Wall Bracket
Veggkrok
Magnetic wall bracket is a convenient alternative way to install the uvex cleaning station.
  • Four magnetic wall brackets provided
  • These brackets give ease of access and allow wearers to keep safety eyewear clean just about anywhere