UV Incubator, benchtop, SI-950

Leverandør: UVP ULTRA VIOLET PRODUCTS
95-0358-02
UVPA95-0358-02EA 45200 NOK
UVPA95-0358-02
UV Incubator, benchtop, SI-950
Inkubatorer Inkubatorer, bordmodell
This benchtop UV Incubator with stainless steel interior is designed to provide precise temperature control and uniformity throughout the chamber. Used for incubation of biological assays, fungal, bacterial cultures, eggs and other biological samples.

  • Microprocessor controlled temperature
  • View samples without opening the door
  • Temperature: 68 °C maximum
  • Shelves: adjustable at three positions
  • Overhead shortwave 254 nm ultraviolet for sterilisating the chamber between experiments

Leveringsinformasjon: Supplied with two ventilated shelves.
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR