Supelco® Cerilliant® Organic Reference Standard, Nicotinamide (Vitamin B3) solution

Leverandør: Merck

V-016-1ML
V-016-1MLEA 830 NOK
V-016-1ML
Supelco® Cerilliant® Organic Reference Standard, Nicotinamide (Vitamin B3) solution
Standarder Organiske standarder

  • GHS signal: GHS02, GHS06, GHS08


NB: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bab9437b0d06449404dd93b4bb46ab11e22e1dd&vid=1bab9437b0d06449404dd93b4bb46ab11e22e1dd&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) Nicotinamide; Vitamin B₃; Niacinamide; Nicotinic acid amide; Pyridine-3-carboxylic acid amide; Vitamin B₃; Vitamin PP
Environmentally Preferable
M-Clarity quality level MQ300
CAS-nr 98-92-0
MDL No. MFCD00006395
Molekylvekt 122,12
Smeltepunkt 128 - 131 °C (lit.)
Hardhetsklasse beskrivelse H225 - H301 + H311 + H331 - H370
Flammepunkt 9,7 °C - Closed cup
Fraktforhold STD
Lagringsforhold –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR