Solid phase extraction cartridges, Bond Elut™ PCB
Leverandør: VARIAN
12105032
VARI12105032EA 12950 NOK
VARI12105032
Solid phase extraction cartridges, Bond Elut™ PCB
Kromatografikolonner
Bond Elut™ PCB is a specially designed sorbent which allows for the facile extraction of polychlorinated biphenyl (PCB) compounds from a variety of matrices. Desired analytes can be loaded and eluted using a simple, single solvent method prior to analysis by GC/ECD.
  • Optimised bed mass affording excellent extraction reproducibility
  • Special dual-phase enhances PCB selectivity
  • All extractions can be completed with one solvent to simplify procedures
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Overflate areal 500 m²/g