Test tubes for autosamplers
Leverandør: VARIAN
3710051100
VARI3710051100EA 1400 NOK
VARI3710051100
Test tubes for autosamplers
Rør Reagensrør
PP, translucent.
  • Without rim
  • Excellent chemical resistance
  • Good temperature resistance