Deactivated injector liners
Leverandør: VARIAN
CP14988
VARICP14988EA 38700 NOK
VARICP14988
Deactivated injector liners
Kromatografiutstyr Kromatografiledninger
These deactivated injector liners are optimised for sensitive and reproducible analysis.
  • Easy-opening storage trays
  • High performance standards
  • Silanisation activation prevents sample degradation