PLOT columns, CP-PoraPLOT Q and CP-PoraPLOT Q-HT
Leverandør: VARIAN
CP7554 CP7551 CP7553 CP7558 CP7559 CP6953 CP7557 CP7549 CP7550 CP7552 CP7555 CP7548 CP7550I5 CP7551I5 CP7554I5 CP7556 CP6954
VARICP7554EA 24600 NOK
VARICP7554 VARICP7551 VARICP7553 VARICP7558 VARICP7559 VARICP6953 VARICP7557 VARICP7549 VARICP7550 VARICP7552 VARICP7555 VARICP7548 VARICP7550I5 VARICP7551I5 VARICP7554I5 VARICP7556 VARICP6954
PLOT columns, CP-PoraPLOT Q and CP-PoraPLOT Q-HT
Kromatografikolonner
CP-PoraPLOT Q is recommended for column switching systems that analyze polar and apolar volatile compounds.
  • Water elutes as a sharp peak and can therefore be quantified, improving productivity
  • Repeatable retention times for long-term stability that enhances efficiency
CP-PoraPLOT Q-HT is the high temperature version of CP-PoraPLOT Q, offering the same benefits but operating up to 290 °C.