Bulk media, PLRP-S
Leverandør: VARIAN
PL1412-6801 PL1412-6K00 PL1412-6K01 PL1412-6K02 PL1412-2101 PL1412-2102 PL1412-2103 PL1412-2200 PL1412-2201 PL1412-2400 PL1412-2401 PL1412-2702 PL1412-2703 PL1412-4100 PL1412-4101 PL1412-4102 PL1412-4103 PL1412-4200 PL1412-4201 PL1412-4400 PL1412-4401 PL1412-4702 PL1412-4703 PL1412-4K00 PL1412-4K01 PL1412-4K02 PL1412-6100 PL1412-6101 PL1412-6102 PL1412-6103 PL1412-6200 PL1412-6201 PL1412-6400 PL1412-6401 PL1412-6702 PL1412-6703 PL1412-6800
VARIPL1412-6801EA 513500 NOK
VARIPL1412-6801 VARIPL1412-6K00 VARIPL1412-6K01 VARIPL1412-6K02 VARIPL1412-2101 VARIPL1412-2102 VARIPL1412-2103 VARIPL1412-2200 VARIPL1412-2201 VARIPL1412-2400 VARIPL1412-2401 VARIPL1412-2702 VARIPL1412-2703 VARIPL1412-4100 VARIPL1412-4101 VARIPL1412-4102 VARIPL1412-4103 VARIPL1412-4200 VARIPL1412-4201 VARIPL1412-4400 VARIPL1412-4401 VARIPL1412-4702 VARIPL1412-4703 VARIPL1412-4K00 VARIPL1412-4K01 VARIPL1412-4K02 VARIPL1412-6100 VARIPL1412-6101 VARIPL1412-6102 VARIPL1412-6103 VARIPL1412-6200 VARIPL1412-6201 VARIPL1412-6400 VARIPL1412-6401 VARIPL1412-6702 VARIPL1412-6703 VARIPL1412-6800
Bulk media, PLRP-S
Kromatografiabsorbenter HPLC sorbenter
The PLRP-S reversed phase HPLC media delivers the pure oligonucleotides (oligos) required for genomics applications, including quantitative PCR, microarrays, and gene synthesis assays.
  • Exceptional resolution and selectivity
  • High purity levels
  • Excellent column lifetime
It easily accommodates high temperatures for better selectivity and resolution between impurities and the target oligo. With PLRP-S stable polymeric reversed phase material, oligos can be purified at temperatures up to 80 °C, while maintaining packed bed integrity.