Chromatography paper
Leverandør: VARIAN
SGI0001 A120B12
VARISGI0001EA 22700 NOK
VARISGI0001 VARIA120B12
Chromatography paper
Filter Filterpapir Kromatografipapir
Chromatography paper is used in thin layer chromatography applications such as evaluating radioisotope purity, QC testing.
  • The porous paper is made of glass microfibers impregnated with silica gel
  • No interference from organic binders
  • Separates lipids and other non-polar compounds
  • Can easily be cut to convenient testing sizes, and can be imprinted
Bestillingsinformasjon: Offers two kinds of paper such as SA (contains sodium salt) and SG (contains potassium salt).