Double Rod Mini H-Stand Set
Leverandør: VELP SCIENTIFIC
A00000484
VELPA00000484EA 8125 NOK
VELPA00000484
Double Rod Mini H-Stand Set
Stativer
The Mini H-stand set is a complete system with a base, double rod, clamp and a vessel holder with safety belt. It is designed to ensure maximum stability, precision and ease of positioning of the sample to be processed.
  • Enhanced stability and reduced vibrations through robust rods
  • Precision-engineered ergonomic knobs minimise operator fatigue and ensure consistent tightening
  • Efficient height adjustment is facilitated by an innovative easy-sliding technology system, enabling swift and precise adjustments
  • Reliability and simplified maintenance are ensured by